Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Protein Synonyms:RAET1G|Retinoic acid early transcript 1G protein
- Protein/Peptide Name:ULBP5
- Purity:>95%
- Sequence:MKHHHHHHAS GLADPHSLCY DITVIPKFRP GPRWCAVQGQ VDEKTFLHYD CGSKTVTPVS PLGKKLNVTT AWKAQNPVLR EVVDILTEQL LDIQLENYIP KEPLTLQARM SCEQKAEGHG SGSWQLSFDG QIFLLFDSEN RMWTTVHPGA RKMKEKWEND KDMTMSFHYI SMGDCTGWLE DFLMGMDSTL EPSAGGTV
- Cat. No.:101189-684
- Supplier no.:RD172315100
ULBP5 acts as a ligand for the NKG2D receptor on the surface of NK cells. ULBP ligands are produced by virally infected cells and tumours. Stimulation of NKG2D contributes to anti-tumour and anti-viral immune responses of NK cells. ULBP5 is expressed in two variants, as a membrane and soluble isoform.
Sequence: MKHHHHHHAS GLADPHSLCY DITVIPKFRP GPRWCAVQGQ VDEKTFLHYD CGSKTVTPVS PLGKKLNVTT AWKAQNPVLR EVVDILTEQL LDIQLENYIP KEPLTLQARM SCEQKAEGHG SGSWQLSFDG QIFLLFDSEN RMWTTVHPGA RKMKEKWEND KDMTMSFHYI SMGDCTGWLE DFLMGMDSTL EPSAGGTV
Other names: Retinoic acid early transcript 1G protein, RAET1G