Order Entry
United States
ContactUsLinkComponent
Angiopoietin-Like Protein 2 Human E. coli, 0.1 mg
Angiopoietin-Like Protein 2 Human E. coli, 0.1 mg
Catalog # 101189-678
Supplier:  BioVendor
CAS Number:  
undefined
Angiopoietin-Like Protein 2 Human E. coli, 0.1 mg
Catalog # 101189-678
Supplier:  BioVendor
Supplier Number:  RD172305100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.1 mg
  • Storage Conditions:
    Store the lyophilized protein at –80 °C. Lyophilized protein remains stable until the expiry date when stored at –80 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at –80 °C for long term storage. Reconstituted protein can be stored at 4 °C for a week
  • Endotoxin Content:
    <0.1 EU/μg
  • Reconstitution Instructions:
    Add 0.1 M Acetate buffer pH 4 to prepare a working stock solution of approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to a concentration of 10 μg/ml. In higher concentrations the solubility of this antigen is limited. Product is not sterile! Please filter the product by an appropriate sterile filter before using it in the cell culture.
  • Protein Synonyms:
    UNQ170/PRO196|ARP2|Angiopoietin-Related Protein 2|ANGPTL2
  • Protein/Peptide Name:
    Angiopoietin-Like Protein 2 Human E. coli
  • Purity:
    >95%
  • Molecular Weight:
    56 kDa (calculated)
  • Sequence:
    MKHHHHHHASQEDGFEGTEEGSPREFIYLNRYKRAGESQDKCTYTFIVPQQRVTGAICVNSKEPEVLLENRVHKQELELLNNELLKQKRQIETLQQLVEVDGGIVSEVKLLRKESRNMNSRVTQLYMQLLHEIIRKRDNALELSQLENRILNQTADMLQLASKYKDLEHKYQHLATLAHNQSEIIAQLEEHCQRVPSARPVPQPPPAAPPRVYQPPTYNRIINQISTNEIQSDQNLKVLPPPLPTMPTLTSLPSSTDKPSGPWRDCLQALEDGHDTSSIYLVKPENTNRLMQVWCDQRHDPGGWTVIQRRLDGSVNFFRNWETYKQGFGNIDGEYWLGLENIYWLTNQGNYKLLVTMEDWSGRKVFAEYASFRLEPESEYYKLRLGRYHGNAGDSFTWHNGKQFTTLDRDHDVYTGNCAHYQKGGWWYNACAHSNLNGVWYRGGHYRSRYQDGVYWAEFRGGSYSLKKVVMMIRPNPNTFH
  • Endotoxin Level:
    <0.1 EU/μg
  • Formulation:
    Filtered (0.4 μm) and lyophilized in 0.5 mg/ml in 0.05 M Acetate buffer pH 4.0
  • Shipping Temperature:
    At ambient temperature
  • Tested Applications:
    Western blotting, ELISA
  • Cat. No.:
    101189-678
  • Supplier no.:
    RD172305100

Specifications

About this item

Total 481 AA. MW: 56 kDa (calculated). UniProtKB acc.no. Q9UKU9 (Gln23-His493). N-terminal His-tag (10 extra AA). Protein identity confirmed by LC-MS/MS.

  • Quality Control Test
  • BCA to determine quantity of the protein
  • SDS PAGE to determine purity of the protein
  • LAL to determine quantity of endotoxin

Sequence: MKHHHHHHAS QEDGFEGTEE GSPREFIYLN RYKRAGESQD KCTYTFIVPQ QRVTGAICVN SKEPEVLLEN RVHKQELELL NNELLKQKRQ IETLQQLVEV DGGIVSEVKL LRKESRNMNS RVTQLYMQLL HEIIRKRDNA LELSQLENRI LNQTADMLQL ASKYKDLEHK YQHLATLAHN QSEIIAQLEE HCQRVPSARP VPQPPPAAPP RVYQPPTYNR IINQISTNEI QSDQNLKVLP PPLPTMPTLT SLPSSTDKPS GPWRDCLQAL EDGHDTSSIY LVKPENTNRL MQVWCDQRHD PGGWTVIQRR LDGSVNFFRN WETYKQGFGN IDGEYWLGLE NIYWLTNQGN YKLLVTMEDW SGRKVFAEYA SFRLEPESEY YKLRLGRYHG NAGDSFTWHN GKQFTTLDRD HDVYTGNCAH YQKGGWWYNA CAHSNLNGVW YRGGHYRSRY QDGVYWAEFR GGSYSLKKVV MMIRPNPNTF H

Other names: Angiopoietin-Related Protein 2, ANGPTL2, ARP2, UNQ170/PRO196

Caution: Research use only.