Order Entry
United States
ContactUsLinkComponent
Human Hsp70 1A/1B (from E. coli)
Human Hsp70 1A/1B (from E. coli)
Catalog # 102552-944
Supplier:  BioVendor
CAS Number:  
undefined
Human Hsp70 1A/1B (from E. coli)
Catalog # 102552-944
Supplier:  BioVendor
Supplier Number:  RD172271100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    HSP70.1/HSP70.2|HSPA1|HSPA1A|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1/2
  • Protein/Peptide Name:
    Hsp70 1A/1B
  • Cat. No.:
    102552-944
  • Supplier no.:
    RD172271100

Specifications

About this item

The Heat shock protein 70(HSP70) family was found in many intracellular compartments. Members of this protein occur in chloroplasts, endoplasmic reticulum, mitochondria, and cytosol. These proteins are induced by a variety of biological stresses, including heat stress, in every organism. HSP70 serves a variety of roles: It acts as molecular chaperones facilitating the assembly of multi-protein complexes, It participates in the translocation of polypeptides across cell membranes and to the nucleus It aids in the proper folding of nascent polypeptide chains. HSP70 is mitochondrial import machinery and plays key roles in the cytosolic endoplasmic reticulum. Recently, extracellular localized HSP have been found to play key roles in the induction of a cellular immune response.

Sequence: MKHHHHHHAS AKAAAIGIDL GTTYSCVGVF QHGKVEIIAN DQGNRTTPSY VAFTDTERLI GDAAKNQVAL NPQNTVFDAK RLIGRKFGDP VVQSDMKHWP FQVINDGDKP KVQVSYKGET KAFYPEEISS MVLTKMKEIA EAYLGYPVTN AVITVPAYFN DSQRQATKDA GVIAGLNVLR IINEPTAAAI AYGLDRTGKG ERNVLIFDLG GGTFDVSILT IDDGIFEVKA TAGDTHLGGE DFDNRLVNHF VEEFKRKHKK DISQNKRAVR RLRTACERAK RTLSSSTQAS LEIDSLFEGI DFYTSITRAR FEELCSDLFR STLEPVEKAL RDAKLDKAQI HDLVLVGGST RIPKVQKLLQ DFFNGRDLNK SINPDEAVAY GAAVQAAILM GDKSENVQDL LLLDVAPLSL GLETAGGVMT ALIKRNSTIP TKQTQIFTTY SDNQPGVLIQ VYEGERAMTK DNNLLGRFEL SGIPPAPRGV PQIEVTFDID ANGILNVTAT DKSTGKANKI TITNDKGRLS KEEIERMVQE AEKYKAEDEV QRERVSAKNA LESYAFNMKS AVEDEGLKGK ISEADKKKVL DKCQEVISWL DANTLAEKDE FEHKRKELEQ VCNPIISGLY QGAGGPGPGG FGAQGPKGGS GSGPTIEEVD

Other names: Heat shock 70 kDa protein 1/2, HSP70-1/HSP70-2, HSP70.1/HSP70.2, HSPA1A, HSPA1, HSPA1B

Ordering information: For research use only.