Order Entry
United States
ContactUsLinkComponent
Human Recombinant Collectrin (from E. coli)
Human Recombinant Collectrin (from E. coli)
Catalog # 101169-494
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Collectrin (from E. coli)
Catalog # 101169-494
Supplier:  BioVendor
Supplier Number:  RD172266100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    UNQ679/PRO1312|TMEM27
  • Protein/Peptide Name:
    Collectrin
  • Purity:
    > 90 %
  • Sequence:
    MKHHHHHHAS ELCQPGAENA FKVRLSIRTA LGDKAYAWDT NEEYLFKAMV AFSMRKVPNR EATEISHVLL CNVTQRVSFW FVVTDPSKNH TLPAVEVQSA IRMNKNRINN AFFLNDQTLE FLKIPSTLAP PMDPSVP
  • Cat. No.:
    101169-494
  • Supplier no.:
    RD172266100

Specifications

About this item

Collectrin regulates insulin secretion on the surface of pancreatic beta cells, and mediates amino acid transport on renal tubular epithelium. The extracellular domain is cleaved off the surface of beta cells.

Sequence: MKHHHHHHAS ELCQPGAENA FKVRLSIRTA LGDKAYAWDT NEEYLFKAMV AFSMRKVPNR EATEISHVLL CNVTQRVSFW FVVTDPSKNH TLPAVEVQSA IRMNKNRINN AFFLNDQTLE FLKIPSTLAP PMDPSVP

Other names: TMEM27, UNQ679/PRO1312

Ordering information: For research use only.