Order Entry
United States
ContactUsLinkComponent
Human Recombinant Cytl1 (from E. coli)
Human Recombinant Cytl1 (from E. coli)
Catalog # 101169-492
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Cytl1 (from E. coli)
Catalog # 101169-492
Supplier:  BioVendor
Supplier Number:  RD172265100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    CYTL1|C4orf4|UNQ1942/PRO4425
  • Protein/Peptide Name:
    Cytl1
  • Purity:
    >95%
  • Sequence:
    MKHHHHHHAS TPPTCYSRMR ALSQEITRDF NLLQVSEPSE PCVRYLPRLY LDIHNYCVLD KLRDFVASPP CWKVAQVDSL KDKARKLYTI MNSFCRRDLV FLLDDCNALE YPIPVTTVLP DRQR
  • Cat. No.:
    101169-492
  • Supplier no.:
    RD172265100

Specifications

About this item

Cytokine-like protein 1 is a secreted protein. It is expressed in CD34+ cell populations of bone marrow and cord blood which function as hematopoietic stem/progenitor cells. It is not expressed in CD34– cells. Experiments with knock-out mice (Cytl1-/-) suggest that CYTL1 is required for the maintenance of cartilage homeostasis rather than cartilage and bone development, and loss of CYTL1 function is associated with experimental osteoarthritic cartilage destruction in mice.

Sequence: MKHHHHHHAS TPPTCYSRMR ALSQEITRDF NLLQVSEPSE PCVRYLPRLY LDIHNYCVLD KLRDFVASPP CWKVAQVDSL KDKARKLYTI MNSFCRRDLV FLLDDCNALE YPIPVTTVLP DRQR

Other names: CYTL1, C4orf4, UNQ1942/PRO4425

Ordering information: For research use only.