Order Entry
United States
ContactUsLinkComponent
Human TFF3 (from E. coli)
Human TFF3 (from E. coli)
Catalog # 102552-836
Supplier:  BioVendor
CAS Number:  
undefined
Human TFF3 (from E. coli)
Catalog # 102552-836
Supplier:  BioVendor
Supplier Number:  RD172160100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    TFF3|ITF|Polypeptide P1.B|TFI|Intestinal trefoil factor|P1.B
  • Protein/Peptide Name:
    TFF3
  • Molecular Weight:
    6.7 kDa
  • Sequence:
    MKHHHHHHAS EEYVGLSANQ CAVPAKDRVD CGYPHVTPKE CNNRGCCFDS RIPGVPWCFK PLQEAECTF
  • Cat. No.:
    102552-836
  • Supplier no.:
    RD172160100

Specifications

About this item

Trefoil factor 3 (TFF3, also known as intestinal trefoil factor) belongs together with TFF1 and TFF2 to a small group of mucin-associated peptides. TFF3 contains seven cysteine residues, six of which form disulfide bonds to create a characteristic three-leafed structure. Due to its compact structure, TFF3 is extremely resistant toward acids, proteolytical cleavage or heat degradation. Monomeric form of TFF3 consists of 60 amino acids and has 6.7 kDa, while the dimer (13.1 kDa) consists of 118 amino acids. TFF3 is expressed mainly in gastrointestinal tract, in the mucous cells of the small and large intestine, where it maintains the integrity of mucous layer and in cooperation with mucins protects the gastrointestinal epithelial cells against various injurious agents. However, TFF3 was also detected in salivary glands, posterior pituitary gland and in the inner ear. Secretion of TFF3 is triggered by the presence of certain inflammation mediators and neurotransmitters.

Sequence: MKHHHHHHAS EEYVGLSANQ CAVPAKDRVD CGYPHVTPKE CNNRGCCFDS RIPGVPWCFK PLQEAECTF

Other names: TFF3, Intestinal trefoil factor, P1.B, ITF, Polypeptide P1.B, TFI

Ordering information: For research use only.