Order Entry
United States
ContactUsLinkComponent
Human Recombinant TFF2 (from E. coli)
Human Recombinant TFF2 (from E. coli)
Catalog # 102552-834
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant TFF2 (from E. coli)
Catalog # 102552-834
Supplier:  BioVendor
Supplier Number:  RD172159100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Gene ID:
    7032
  • Protein Synonyms:
    SP|SML1|TFF2|Spasmolytic polypeptide|Spasmolysin
  • Protein/Peptide Name:
    TFF2
  • Molecular Weight:
    12 kDa
  • Sequence:
    MKHHHHHHAS EKPSPCQCSR LSPHNRTNCG FPGITSDQCF DNGCCFDSSV TGVPWCFHPL PKQESDQCVM EVSDRRNCGY PGISPEECAS RKCCFSNFIF EVPWCFFPKS VEDCHY
  • Cat. No.:
    102552-834
  • Supplier no.:
    RD172159100

Specifications

About this item

Trefoil factor 2 (TFF2) (PSP – pancreatic spasmolytic polypeptide) is a small secreted protein with a molecular weight of 12 kDa. It belongs to the TFF protein family that is characterized by a clover leaf–like disulphide structure named the TFF domain, which is created by 6 cysteines forming three intramolecular bonds. TFF2 contains two trefoil domains and has a very compact structure which may account for its extremely high resistance against the harsh environment in the gastrointestinal tract. The most abundant expression of TFFs is found in GI tract (especially in stomach, colon and pancreas), where they are co-localised with mucins. TFF2 is usually co-localised with MUC6 and probably mediates mucin cross-linking and stabilization of the mucin layer as do the other members of the TFF family.

Sequence: MKHHHHHHAS EKPSPCQCSR LSPHNRTNCG FPGITSDQCF DNGCCFDSSV TGVPWCFHPL PKQESDQCVM EVSDRRNCGY PGISPEECAS RKCCFSNFIF EVPWCFFPKS VEDCHY

Other names: Spasmolytic polypeptide, SP, Spasmolysin, TFF2, SML1

Ordering information: For research use only.