Order Entry
United States
ContactUsLinkComponent
Human CTRP6 (from E. coli)
Human CTRP6 (from E. coli)
Catalog # 102552-828
Supplier:  BioVendor
CAS Number:  
undefined
Human CTRP6 (from E. coli)
Catalog # 102552-828
Supplier:  BioVendor
Supplier Number:  RD172156100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    CTRP6|UNQ581/PRO1151|C1QTNF6
  • Protein/Peptide Name:
    CTRP6
  • Molecular Weight:
    27.02 kDa
  • Sequence:
    MKHHHHHHAS TFDRAVASGC QRCCDSEDPL DPAHVSSASS SGRPHALPEI RPYINITILK GDKGDPGPMG LPGYMGREGP QGEPGPQGSK GDKGEMGSPG APCQKRFFAF SVGRKTALHS GEDFQTLLFE RVFVNLDGCF DMATGQFAAP LRGIYFFSLN VHSWNYKETY VHIMHNQKEA VILYAQPSER SIMQSQSVML DLAYGDRVWV RLFKRQRENA IYSNDFDTYI TFSGHLIKAE DD
  • Cat. No.:
    102552-828
  • Supplier no.:
    RD172156100

Specifications

About this item

Total 242 AA. MW: 27.02 kDa (calculated). UniProtKB acc.no. Q9BXI9. N-terminal His-tag, 10extra AA

Sequence: MKHHHHHHAS TFDRAVASGC QRCCDSEDPL DPAHVSSASS SGRPHALPEI RPYINITILK GDKGDPGPMG LPGYMGREGP QGEPGPQGSK GDKGEMGSPG APCQKRFFAF SVGRKTALHS GEDFQTLLFE RVFVNLDGCF DMATGQFAAP LRGIYFFSLN VHSWNYKETY VHIMHNQKEA VILYAQPSER SIMQSQSVML DLAYGDRVWV RLFKRQRENA IYSNDFDTYI TFSGHLIKAE DD

Other names: C1QTNF6, CTRP6, UNQ581/PRO1151

Ordering information: For research use only.