Human Recombinant CTRP2 (from E. coli)
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Total 280 AA. MW: 29 640 Da (calculated). UniProtKB acc.no. Q9BXJ5. N-Terminal His-tag, 10 extra AA
Sequence: MKHHHHHHAS DPLLGAFARR DFRKGSPQLV CSLPGPQGPP GPPGAPGPSG MMGRMGFPGK DGQDGHDGDR GDSGEEGPPG RTGNRGKPGP KGKAGAIGRA GPRGPKGVNG TPGKHGTPGK KGPKGKKGEP GLPGPCSCGS GHTKSAFSVA VTKSYPRERL PIKFDKILMN EGGHYNASSG KFVCGVPGIY YFTYDITLAN KHLAIGLVHN GQYRIRTFDA NTGNHDVASG STILALKQGD EVWLQIFYSE QNGLFYDPYW TDSLFTGFLI YADQDDPNEV
Other names: C1QTNF2, CTRP2, UNQ6349/PRO21054
Ordering information: For research use only.
Specifications
Specifications
- Size:100 µg
- Cat. No.:101189-782
- Protein/Peptide Type:Recombinant
- Protein Synonyms:C1QTNF2|UNQ6349/PRO21054|CTRP2
- Protein/Peptide Name:CTRP2
- Sequence:MKHHHHHHAS DPLLGAFARR DFRKGSPQLV CSLPGPQGPP GPPGAPGPSG MMGRMGFPGK DGQDGHDGDR GDSGEEGPPG RTGNRGKPGP KGKAGAIGRA GPRGPKGVNG TPGKHGTPGK KGPKGKKGEP GLPGPCSCGS GHTKSAFSVA VTKSYPRERL PIKFDKILMN EGGHYNASSG KFVCGVPGIY YFTYDITLAN KHLAIGLVHN GQYRIRTFDA NTGNHDVASG STILALKQGD EVWLQIFYSE QNGLFYDPYW TDSLFTGFLI YADQDDPNEV
- Species:Human
- Source:E. coli
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



