Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Source:HEK293 cells
- Species:Human
- Size:25 µg
- Gene ID:8742
- Protein Synonyms:APO3L|TNFSF12|TNF-related weak inducer of apoptosis|APO3 ligand|TWEAK|DR3LG
- Protein/Peptide Name:TWEAK
- Molecular Weight:18.4 kDa
- Sequence:ASSAPKGRKT RARRAIAAHY EVHPRPGQDG AQAGVDGTVS GWEEARINSS SPLRYNRQIG EFIVTRAGLY YLYCQVHFDE GKAVYLKLDL LVDGVLALRC LEEFSATAAS SLGPQLRLCQ VSGLLALRPG SSLRIRTLPW AHLKAAPFLT YFGLFQVHKL HHHHHH
- Cat. No.:102552-822
- Supplier no.:RD172148100
Total 166 AA. MW: 18.4 kDa (calculated). UniProtKB acc. No. O43508 (Ser94–His249). N-terminal linker (2 extra AA), C-terminal linker (2 extra AA) and C-terminal His-tag (6 extra AA). Protein identity confirmed by LC-MS/MS.
Sequence: ASSAPKGRKT RARRAIAAHY EVHPRPGQDG AQAGVDGTVS GWEEARINSS SPLRYNRQIG EFIVTRAGLY YLYCQVHFDE GKAVYLKLDL LVDGVLALRC LEEFSATAAS SLGPQLRLCQ VSGLLALRPG SSLRIRTLPW AHLKAAPFLT YFGLFQVHKL HHHHHH
Other names: APO3 ligand; TNF-related weak inducer of apoptosis; TWEAK; TNFSF12; APO3L, DR3LG