Order Entry
United States
ContactUsLinkComponent
Human Recombinant ENPP1 (from HEK293 cells)
Human Recombinant ENPP1 (from HEK293 cells)
Catalog # 101169-484
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant ENPP1 (from HEK293 cells)
Catalog # 101169-484
Supplier:  BioVendor
Supplier Number:  RD172124100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    Phosphodiesterase I/nucleotide pyrophosphatase 1|NPP-1|Ectonucleotide pyrophosphatase/phosphodiesterase family member 1|PC-1|E-NPP 1|Membrane component chromosome 6 surface marker 1|Plasma-cell membrane glycoprotein
  • Protein/Peptide Name:
    ENPP1
  • Molecular Weight:
    96.5 kDa
  • Cat. No.:
    101169-484
  • Supplier no.:
    RD172124100

Specifications

About this item

Total 840 AA, UniProt P22413 (Lys98-Asp925). MW: 96.5 kDa (calculated), migrates at ~ 110 kDa on SDS PAGE; N-terminal linker (2 extra AA), C-terminal linker (4 extra AA) and C-terminal His-tag (6 extra AA). Protein identity confirmed by LC-MS/MS.

Sequence: ASKPSCAKEV KSCKGRCFER TFGNCRCDAA CVELGNCCLD YQETCIEPEH IWTCNKFRCG EKRLTRSLCA CSDDCKDKGD CCINYSSVCQ GEKSWVEEPC ESINEPQCPA GFETPPTLLF SLDGFRAEYL HTWGGLLPVI SKLKKCGTYT KNMRPVYPTK TFPNHYSIVT GLYPESHGII DNKMYDPKMN ASFSLKSKEK FNPEWYKGEP IWVTAKYQGL KSGTFFWPGS DVEINGIFPD IYKMYNGSVP FEERILAVLQ WLQLPKDERP HFYTLYLEEP DSSGHSYGPV SSEVIKALQR VDGMVGMLMD GLKELNLHRC LNLILISDHG MEQGSCKKYI YLNKYLGDVK NIKVIYGPAA RLRPSDVPDK YYSFNYEGIA RNLSCREPNQ HFKPYLKHFL PKRLHFAKSD RIEPLTFYLD PQWQLALNPS ERKYCGSGFH GSDNVFSNMQ ALFVGYGPGF KHGIEADTFE NIEVYNLMCD LLNLTPAPNN GTHGSLNHLL KNPVYTPKHP KEVHPLVQCP FTRNPRDNLG CSCNPSILPI EDFQTQFNLT VAEEKIIKHE TLPYGRPRVL QKENTICLLS QHQFMSGYSQ DILMPLWTSY TVDRNDSFST EDFSNCLYQD FRIPLSPVHK CSFYKNNTKV SYGFLSPPQL NKNSSGIYSE ALLTTNIVPM YQSFQVIWRY FHDTLLRKYA EERNGVNVVS GPVFDFDYDG RCDSLENLRQ KRRVIRNQEI LIPTHFFIVL TSCKDTSQTP LHCENLDTLA FILPHRTDNS ESCVHGKHDS SWVEELLMLH RARITDVEHI TGLSFYQQRK EPVSDILKLK THLPTFSQED GPKLHHHHHH

Other names: Ectonucleotide pyrophosphatase/phosphodiesterase family member 1, E-NPP 1, Membrane component chromosome 6 surface marker 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, NPP-1, Plasma-cell membrane glycoprotein, PC-1

Ordering information: For research use only.