Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Tag sequence:His-tag
- Protein Synonyms:Prostaglandin-H2 D-isomerase|Glutathione-independent PGD synthetase|Lipocalin-Type Prostaglandin D Synthase|Beta-trace protein|PGDS2|BTP|PGD2 synthase|PGDS|PTGDS|Lipocalin-type prostaglandin-D synthase|PDS|Cerebrin-28|Prostaglandin-D2 synthase|EC=5.3.99.2
- Protein/Peptide Name:BTP
- Sequence:MRGSHHHHHH GMASAPEAQV SVQPNFQQDK FLGRWFSAGL ASNSSWLREK KAALSMCKSV VAPATDGGLN LTSTFLRKNQ CETRTMLLQP AGSLGSYSYR SPHWGSTYSV SVVETDYDQY ALLYSQGSKG PGEDFRMATL YSRTQTPRAE LKEKFTAFCK AQGFTEDTIV FLPQTDKCMT EQ
- Cat. No.:200068-180
- Supplier no.:RD172113100
Specifications
About this item
BTP catalyzes the conversion of PGH2 to PGD2, a prostaglandin involved in smooth muscle contraction/relaxation and a potent inhibitor of platelet aggregation. Involved in a variety of CNS functions, such as sedation, NREM sleep and PGE2-induced allodynia, and may have an anti-apoptotic role in oligodendrocytes. Binds small non-substrate lipophilic molecules, including biliverdin, bilirubin, retinal, retinoic acid and thyroid hormone, and may act as a scavenger for harmful hydrophopic molecules and as a secretory retinoid and thyroid hormone transporter. Possibly involved in development and maintenance of the blood-brain, blood-retina, blood-aqueous humor and blood-testis barrier. It is likely to play important roles in both maturation and maintenance of the central nervous system and male reproductive system.
Sequence: MRGSHHHHHH GMASAPEAQV SVQPNFQQDK FLGRWFSAGL ASNSSWLREK KAALSMCKSV VAPATDGGLN LTSTFLRKNQ CETRTMLLQP AGSLGSYSYR SPHWGSTYSV SVVETDYDQY ALLYSQGSKG PGEDFRMATL YSRTQTPRAE LKEKFTAFCK AQGFTEDTIV FLPQTDKCMT EQ
Other names: BTP, Prostaglandin-H2 D-isomerase, Lipocalin-Type Prostaglandin D Synthase, Prostaglandin-H2 D-isomerase, EC=5.3.99.2, Lipocalin-type prostaglandin-D synthase, Glutathione-independent PGD synthetase, Prostaglandin-D2 synthase, PGD2 synthase, PGDS2, PGDS, Beta-trace protein, Cerebrin-28, PTGDS, PDS