Order Entry
United States
ContactUsLinkComponent
Human Rhu Fgf-21 (fromE. coli)
Human Rhu Fgf-21 (fromE. coli)
Catalog # 101422-258
Supplier:  BioVendor
CAS Number:  
undefined
Human Rhu Fgf-21 (fromE. coli)
Catalog # 101422-258
Supplier:  BioVendor
Supplier Number:  RD172108100B
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    FGF-21|UNQ3115/PRO10196
  • Protein/Peptide Name:
    Rhu Fgf-21
  • Molecular Weight:
    ~17-26 kDa
  • Sequence:
    MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS
  • Cat. No.:
    101422-258
  • Supplier no.:
    RD172108100B

Specifications

About this item

The FGFs are a family of more than 20 small (~17–26 kDa) secreted peptides. The initial characterization of these proteins focused on their ability to stimulate fibroblast proliferation. This mitogenic activity was mediated through FGF receptors (FGFRs) 1, 2, or 3. A fourth closely related tyrosine kinase receptor (FGFR4) was able to bind the FGFs but did not lead to a mitogenic response.

Sequence: MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS

Other names: FGF-21, UNQ3115/PRO10196

Ordering information: For research use only.