Order Entry
United States
ContactUsLinkComponent
Human Regenerating I (fromE. coli)
Human Regenerating I (fromE. coli)
Catalog # 101215-292
Supplier:  BioVendor
CAS Number:  
undefined
Human Regenerating I (fromE. coli)
Catalog # 101215-292
Supplier:  BioVendor
Supplier Number:  RD172083100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    Gastrointestinal secretory protein|Regenerating islet-derived protein 4|Regenerating islet-derived protein IV|REG4|REG-like protein|GISP|RELP|REG-4|Reg IV
  • Protein/Peptide Name:
    Regenerating I
  • Sequence:
    MKHHHHHHAS HMDIIMRPSC APGWFYHKSN CYGYFRKLRN WSDAELECQS YGNGAHLASI LSLKEASTIA EYISGYQRSQ PIWIGLHDPQ KRQQWQWIDG AMYLYRSWSG KSMGGNKHCA EMSSNNNFLT WSSNECNKRQ HFLCKYRP
  • Cat. No.:
    101215-292
  • Supplier no.:
    RD172083100

Specifications

About this item

Regenerating (Reg) gene family belongs to the calcium depending lectin gene super family. It represents a group of small, multi‑functional secreted proteins, which can function as acute phase reactants, lectins and anti‑apoptotic or growth agents. These agents play an important role in proliferation and differentiation in the entire GI tract. The Reg family consists of seven members in mice (Reg I, Reg II, Reg IIIa, Reg IIIb, Reg IIId and Reg IIIge and Reg IV). Four members are recognized in humans (Reg Ia, Reg Ib, Reg III and Reg IV), but there are most probably a few more. Reg genes are up‑regulated following tissue injury, and play a major role in the healing of gastrointestinal mucosal lesions. Different members of the Reg gene family were shown to be expressed in pancreatic, gastric and colorectal cancers, and may serve as markers for poor prognosis. Reg IV, a novel member of the family, was suggested to play an important role in initiating the multi‑step process of colorectal cancer carcinogenesis, at the level of adenoma, by increasing the resistance for programmed cell death. Regenerating gene family member 4 (REG4) was originally identified by sequencing of a cDNA library derived from patients with inflammatory bowel disease.

Sequence: MKHHHHHHAS HMDIIMRPSC APGWFYHKSN CYGYFRKLRN WSDAELECQS YGNGAHLASI LSLKEASTIA EYISGYQRSQ PIWIGLHDPQ KRQQWQWIDG AMYLYRSWSG KSMGGNKHCA EMSSNNNFLT WSSNECNKRQ HFLCKYRP

Other names: Regenerating islet-derived protein 4, REG-4, Regenerating islet-derived protein IV, Reg IV, REG-like protein, Gastrointestinal secretory protein, REG4, GISP, RELP

Ordering information: For research use only.