ContactUsLinkComponent
Calpain Activity Assay Kit, PromoKine, PromoCell
Calpain Activity Assay Kit, PromoKine, PromoCell
  10183-666
 :  Avantor
 :  
Calpain Activity Assay Kit, PromoKine, PromoCell
  10183-666
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Calpain Activity Assay Kit
  • Size:
    100 assays
  • Environmentally Preferable:
  • Cat. No.:
    10183-666
  • Supplier no.:
    PK-CA577-K240
The Calpain Activity Assay Kit provides a simple and convenient means for analyzing calpain activity in apoptotic and other samples.
  • Based on fluorometric detection and quantitation of the cleavage of a calpain substrate
  • Positive and negative controls
  • Determination of changes in calpain activity
Activation of calpain is involved in many physiological and pathological processes (e.g., apoptosis). The Calpain Activity Assay Kit provides a simple and convenient means for analyzing calpain activity in apoptotic and other samples. The assay is based on fluorometric detection and quantitation of the cleavage of a calpain substrate at 505 nm using a fluorometer or a fluorescence plate reader. The kit contains both positive and negative controls. Comparison of the fluorescence intensity from a treated sample with a normal control allows determination of the changes in calpain activity. Active calpain and calpain inhibitors and antibodies are also available separately.