Order Entry
United States
ContactUsLinkComponent
Hydrogen peroxide 6%, Peroxigen
Hydrogen peroxide 6%, Peroxigen
  75779-618
 :  Avantor
 :  
undefined
Hydrogen peroxide 6%, Peroxigen
  75779-618
 :  Avantor
 :  

 

  • Size:
    12 x 473 mL (12 x 16 oz.)
  • Cat. No.:
    75779-618
  • Supplier no.:
    9816

 

 

 :
 :150.2 °C (1013 hPa)
 :–0.41 °C
 :1.11 g/cm³ (20 °C)
 :5.1,II
 :34.01
 :2014
 :7722-84-1
 :H₂O₂
 :MFCD00011333