Luciferase Reporter Assay Kit I (Firefly), PromoKine
Supplier: Avantor
Certificates
About this item
PromoKine's Luciferase Reporter Assay kit is designed for detection and quantification of the Firefly luciferase reporter enzyme from cultured cells in a simple, efficient, and linear fashion.
- Flash-type luminescence assay
- Signal half-life of about 12 minutes
- Detection and quantification
PromoKine's Luciferase Reporter Assay kit is designed for detection and quantification of the Firefly luciferase reporter enzyme from cultured cells in a simple, efficient, and linear fashion. This is a flash-type luminescence assay with signal half-life of about 12 minutes. PromoKine also offers the Luciferase Reporter HTS Kit which is a homogenous glow-type assay with signal half-life of three to five hours.
Frequently Bought Together