ContactUsLinkComponent
SuperFastPrep-2™ Sample Preparation Instrument, MP Biomedicals
SuperFastPrep-2™ Sample Preparation Instrument, MP Biomedicals
  75784-598
undefined
SuperFastPrep-2™ Sample Preparation Instrument, MP Biomedicals
  75784-598
 :  116012500
  • Description:
    SuperFastPrep-2™ Sample Preparation Instrument
  • Cat. No.:
    75784-598
  • Supplier no.:
    116012500
The SuperFastPrep-2™ is an attachment for a rotary tool developed for vigorous cell disruption (bead beating) and allows the researcher/investigator to process samples in the field, at the site of sample collection.
  • Lyse microbes in soil, sediment, sludge, and fecal samples
  • Compatible with most 2 mL tubes containing lysis matrix
  • Ideal for cordless, portable, and convenient processing of tough-to-lyse biological and environmental samples
The SuperFastPrep-2™ is an attachment for a rotary tool developed for vigorous cell disruption (bead beating) and allows the researcher/investigator to process samples in the field, at the site of sample collection
Suitable for all sample types; Lysing highly contagious samples; Lysis of yeast and fungi; Used for Isolation of PCR-Ready Genomic DNA from Fecal Samples; Used for Isolation of PCR-Ready Genomic DNA from Fungi and Bacteria Samples; Used for Isolation of PCR-Ready Genomic DNA from Soil Samples; DNA & RNA Purification; Lysis of bacteria; Lysis of soft tissues; Isolation of genomic DNA; Purification of DNA; Cell Lysis; Used to lyse cell wall of yeast; Break hard/brittle samples; Used to isolate DNA from virtually any source
It can be used to lyse microbes in soil, sediment, sludge, and fecal samples and can effectively process tough-to-lyse fungal, algal, plant, and animal tissues. It can be used at any remote location and in most weather conditions when immediate sample collection, processing, and preservation are required. The device is compatible with most 2 mL tubes containing lysis matrix. The SuperFastPrep-2 consists of a unique, reciprocating grinding mechanism mounted on a rotary tool. It is designed to operate at 600 to 4,800 cycles per minute as long as charge remains in the battery. This allows the researcher to maintain control of the speed and time of sample lysis. The SuperFastPrep-2™ is ideal for cordless, portable, and convenient processing of tough-to-lyse biological and environmental samples, both in the lab and in the field.