PTV Slotted Capillary Column Nut for Gerstel CIS 3 and CIS 4 PTV Inlets, Restek
Catalog # 10855-996
Supplier: Restek
Specifications
- Description:PTV Slotted Capillary Column Nut
- Cat. No.:10855-996
- For use with:for Gerstel CIS 3 and CIS 4 PTV Inlets
- Supplier no.:22349
Specifications
About this item
For use with Gerstel CIS 3 and CIS 4 PTV inlets.