Accessories for Parker HydroGen Mate™ DI Water System, Restek
Catalog # RK22001
Supplier: Restek
Specifications
- Description:Maintenance Kit for Parker HydroGen Mate DI Water System
- Cat. No.:RK22001
- Includes:Prefilter (1); Resin Cartridges (2) and Final Filter (1)
- Supplier no.:22001