Septa for SRI GCs, Plug, Restek
Supplier: Avantor
Certificates
About this item
Restek Thermolite® septa are compatible with differing internal diameters depending on vial size, and functional temperatures differ between models.
- Effective seal
- Easy puncturability
These septa are available in a range of sizes for use with a variety of vials.
Packaging: Packaged in ultra-clean blister packs.
Caution: Note: Due to differences in inlet design, the actual septum temperature for a given inlet setpoint can vary by manufacturer. Restek recommends using only BTO® septa in Thermo TRACE and Focus® GCs. For 17 mm inlets, the maximum temperature is 330 °C. For all injectors, minimum recommended operating temperature for BTO® septa is 250 °C.
Frequently Bought Together