Zero Dead Volume Tee, Valco®, Restek
Supplier: Restek
Certificates
About this item
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
- Fittings are available in brass or stainless steel
- Treated with Siltek®/Sulfinert
Use these tee connectors for making three-way connections with tubing. Available with connection sizes of 1/16" or 1/8".
Frequently Bought Together