Order Entry
United States
ContactUsLinkComponent
PTV Inlet Adapter Kit for Gerstel CIS 3 and CIS 4 PTV Inlets, Restek
PTV Inlet Adapter Kit for Gerstel CIS 3 and CIS 4 PTV Inlets, Restek
Catalog # 10855-998
Supplier:  Restek
undefined
PTV Inlet Adapter Kit for Gerstel CIS 3 and CIS 4 PTV Inlets, Restek
Catalog # 10855-998
Supplier:  Restek
Supplier Number:  22350

Specifications

  • Description:
    PTV Inlet Adaptor Kit for 0.25 and 0.32 mm ID Columns
  • Cat. No.:
    10855-998
  • For use with:
    for Gerstel CIS 3 and CIS 4 PTV Inlets
  • Supplier no.:
    22350

Specifications

About this item

PTV inlet adapter kits are suitable for 0.25 mm and 0.32 mm ID columns.

  • Meets original manufacturer's performance

For use with Gerstel CIS 3 and CIS 4 PTV inlets.

Includes inlet adapter, silver PTV seal, and slotted capillary nut.