ContactUsLinkComponent
Restek Enhanced Capillary Nut for Shimadzu 17A, 2010, and 2014 GCs, Restek
Restek Enhanced Capillary Nut for Shimadzu 17A, 2010, and 2014 GCs, Restek
Catalog # 10058-558
Supplier:  Restek
undefined
Restek Enhanced Capillary Nut for Shimadzu 17A, 2010, and 2014 GCs, Restek
Catalog # 10058-558
Supplier:  Restek
Supplier Number:  20375
Specifications
  • Description:
    Restek Enhanced Capillary Nut
  • Cat. No.:
    10058-558
  • For use with:
    for Shimadzu 17A, 2010, and 2014 GCs
  • Supplier no.:
    20375
Specifications
About this item
Restek's enhanced design for the capillary nut eliminates the slot, increasing lifetime and durability.
  • All parts meet or exceed original manufacturer's performance
For use with Shimadzu 17A, 2010, and 2014 GCs.