ContactUsLinkComponent
Ion Source Cleaning Powder, Restek
Ion Source Cleaning Powder, Restek
Catalog # 89205-676
Supplier:  Restek
undefined
Ion Source Cleaning Powder, Restek
Catalog # 89205-676
Supplier:  Restek
Supplier Number:  22685
Specifications
  • Description:
    Ion Source Cleaning Powder
  • Cat. No.:
    89205-676
  • Supplier no.:
    22685
Specifications
About this item
Use this aluminum oxide powder to clean surfaces that contact the sample or ion beam when you encounter poor sensitivity and inadequate abundances at high masses.