Nut Driver, ¹/₄", Restek
6.4 mm (¼")
Catalog # 10058-596
Supplier: Restek
Supplier #: 21076
Jump to:
Product Details & Documents
Ideal for changing Agilent 5890/6850/6890 FID jets.
- Securely grips jet for easy removal or installation
Specifications
- Bit diameter:6.4 mm (¼")