ContactUsLinkComponent
Dual Column mini-Lam Direct Injection Tee Kit for ¹/₄" packed column inlets, Restek
Dual Column mini-Lam Direct Injection Tee Kit for ¹/₄" packed column inlets, Restek
Catalog # 75778-664
Supplier:  Avantor
undefined
Dual Column mini-Lam Direct Injection Tee Kit for ¹/₄" packed column inlets, Restek
Catalog # 75778-664
Supplier:  Avantor
Specifications
  • Description:
    Mini-Lam DI Tee Kit
  • Cat. No.:
    75778-664
  • Includes:
    All fittings and ferrules
  • Supplier no.:
    20436
Specifications
About this item
Inverted cup design offers complete sample vaporization.
  • Permits larger sample volumes
  • Incorporates a Press-Tight® taper in each outlet leg, to make a perfect, dead-volume-free connection to each analytical column (ODs ranging from 0.4 to 0.8 mm)
  • Allows visual confirmation of the column connection
Open-top design makes it easy to pack with glass wool, keeping sample residue from contaminating the cup.