Indicating Moisture Trap, Restek
: Restek
Indicating moisture traps reduce water to less than 10 ppb; indicator changes from yellowish-green to blue at 5% relative humidity.
- Prepurged for fast stabilization
- Reduces noise from high-sensitivity detectors
- 10 µm frit prevents microparticulate damage to needle valves and flow controllers
- Maximum operating pressure: 100 psi (689 kPa)
The heavy-walled glass body of these traps prevents oxygen and water infusion.
Frequently Bought Together