Order Entry
United States
ContactUsLinkComponent
Vu-Tight DI Liners for ¹/₄" Packed Injection Port Conversion, Restek
Vu-Tight DI Liners for ¹/₄" Packed Injection Port Conversion, Restek
Catalog # RK20343
Supplier:  Restek
undefined
Vu-Tight DI Liners for ¹/₄" Packed Injection Port Conversion, Restek
Catalog # RK20343
Supplier:  Restek
Supplier Number:  20343

Specifications

  • Ext.Ø:
    1/4"
  • Description:
    Vu-Tight DI Liner
  • Cat. No.:
    RK20343
  • For use with:
    Column ODs from 0.5 - 0.8 mm
  • Column I.D.:
    0.32 and 0.53 mm ID Capillary Columns
  • Supplier no.:
    20343

Specifications

About this item

Visually observe the Press-Tight® connection between the column end and liner.

  • Slotted top prevents obstruction of carrier gas flow
  • Operate in the direct injection mode

Can easily be packed with wool for dirty samples.