ContactUsLinkComponent
Replacement Cables for Agilent GCs, Integrators, & Autosamplers, Restek
Replacement Cables for Agilent GCs, Integrators, & Autosamplers, Restek
Catalog # 10855-860
Supplier:  Avantor
Replacement Cables for Agilent GCs, Integrators, & Autosamplers, Restek
Catalog # 10855-860
Supplier:  Avantor
Specifications
  • Description:
    Replacement cable
  • Length:
    1.8 m (6 ft.)
  • Cat. No.:
    10855-860
  • Supplier no.:
    20652
Specifications
About this item
Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder.
  • Instructions and wiring diagrams included
  • Manufactured with only the highest-quality components
  • 6-foot length
For use with Agilent GCs, integrators, and autosamplers.