ContactUsLinkComponent
MXT® Needle Files, Restek
MXT® Needle Files, Restek
  10852-892
 :  Restek
undefined
MXT® Needle Files, Restek
  10852-892
 :  Restek
 :  21601
  • Description:
    Needle Files
  • Cat. No.:
    10852-892
  • Supplier no.:
    21601
Multi-purpose files are especially useful for cutting Siltek®-treated stainless steel columns.
  • Durable stainless steel fabrication