Stainless Steel Jet Reamers/Ferrule Removers, Restek
Catalog # RK20113
Supplier: Restek
Specifications
- Description:Stainless Steel Jet Reamers/Ferrule Removers
- Cat. No.:RK20113
- Supplier no.:20113
Specifications
About this item
Stainless steel jet reamers are great tools for cleaning detector jets or removing stuck ferrules from other small orifices.
- Useful for fine cleaning operations
- Slender design
Serrated design is optimal for removing silica deposits and other contaminants.