ContactUsLinkComponent
Etest® Inoculator Retro C80, bioMerieux
Etest® Inoculator Retro C80, bioMerieux
Catalog # 10500-472
Supplier:  Avantor
Etest® Inoculator Retro C80, bioMerieux
Catalog # 10500-472
Supplier:  Avantor
Specifications
  • Description:
    Inoculator Retro
  • Cat. No.:
    10500-472
  • Supplier no.:
    559803
Specifications
About this item
Retro C80 is a rota-plater that simplifies and standardizes the inoculation of small and large agar plates making Etest easier to read when compared to manual streaking
  • Reduces operator fatigue
  • Makes readings easier
  • Standardizes inoculation
Allows easier application of samples to testing equipment.