Mini Grip-It Device, bioMérieux
Catalog # 10500-304
Supplier: bioMerieux
Specifications
- Description:Mini Grip-It, 100/pack
- Cat. No.:10500-304
- Supplier no.:411200
Specifications
About this item
The Mini Grip-It is a device that makes it easier to grab, hold, and move small and light objects from one place to another.
- Provides greater control of small objects
- Handles lab equipment easily
- Can be autoclaved
The suction cup picks up hard to handle items such as Etest strips, antibiotic disks, and glass/plastic cover slips. The whole apparatus can be autoclaved.