ContactUsLinkComponent
API® M Medium Microbial Identification Kit, bioMerieux
API® M Medium Microbial Identification Kit, bioMerieux
Catalog # 89126-946
Supplier:  bioMerieux
CAS Number:  
API® M Medium Microbial Identification Kit, bioMerieux
Catalog # 89126-946
Supplier:  bioMerieux
Supplier Number:  50120
CAS Number:  
Specifications
  • Description:
    API® M Medium Microbial Identification Kit
  • Cat. No.:
    89126-946
  • Supplier no.:
    50120
Specifications
About this item
Tests for motility of facultative anaerobic bacteria.
  • Economical
  • Easy to use
Motility medium reagents for use with API® 20 strips.