Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL22 (from CHO cells)
Human Recombinant IL22 (from CHO cells)
Catalog # 75794-392
Supplier:  Prosci
CAS Number:  
Human Recombinant IL22 (from CHO cells)
Catalog # 75794-392
Supplier:  Prosci
Supplier Number:  90-515
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    25 μg
  • Tag sequence:
    Fc Tag
  • Storage Conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein Synonyms:
    IL-10-related T Cell-derived-inducible Factor|Interleukin-22
  • Protein/Peptide Name:
    IL22
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    34-179
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Cat. No.:
    75794-392
  • Supplier no.:
    90-515

Specifications

About this item

Interleukin-22 (IL-22), also known as IL-10 related T cell derived inducible factor (ILTIF) was initially identified as a gene induced by IL-9 in mouse T cells and mast cells. IL-22 has been shown to activate STAT1 and STAT3 in several hepatoma cell lines and upregulate the production of acute phase proteins. IL-22 is produced by normal T cells upon anti-CD3 stimulation in humans. Mouse IL-22 expression is also induced in various organs upon lipopolysaccharide injection, suggesting that IL-22 may be involved in inflammatory responses. The functional IL-22 receptor complex consists of two receptor subunits, IL-22R(CRF29) and IL-10Rbeta(CRF24), belonging to the class II cytokine receptor family.

  • Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.