Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant FLT3 Ligand (from CHO cells)
  75794-106
 :  Prosci
 :  
Human Recombinant FLT3 Ligand (from CHO cells)
  75794-106
 :  Prosci
 :  90-300
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    10 μg
  • Tag sequence:
    Fc Tag
  • Storage Conditions:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biological Activity:
    In vitro and in vivo generation of conventional and plasmacytoid dendritic cells (DC).
    FLT3L increases in vivo both NK and DC cells numbers in the spleen when mice are injected intraperitoneally with 2 or 10 ug, daily for 10 days (slightly higher effect at 10 ug).
  • Protein Synonyms:
    Fms-related tyrosine kinase 3 ligand
  • Protein/Peptide Name:
    FLT3 Ligand
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Liquid, In PBS (sterile).
  • Cat. No.:
    75794-106
  • Supplier no.:
    90-300

 

 

FMS-like tyrosine kinase 3 ligand (FLT3L) acts as a growth factor that increases the number of immune cells (lymphocytes (B cells and T cells)) by activating the hematopoietic progenitors. It binds to FLT3 (CD135) which is found on multipotent progenitor (MPP) and common lymphoid progenitor (CLP) cells in mice. FLT3L induces the mobilization of the hematopoietic progenitors and stem cells in vivo which may help the system to kill cancer cells. FLT3L is crucial for steady-state pDC (plasmacytoid dendritic cells) and cDC (classical dendritic cells) development. Deficiency of FLT3L causes a dramatic decrease in DC numbers, whereas increasing its availability increase in DC numbers.

  • Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.