Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant SEMA4C
Mouse Recombinant SEMA4C
Catalog # 75791-506
Supplier:  Prosci
CAS Number:  
Mouse Recombinant SEMA4C
Catalog # 75791-506
Supplier:  Prosci
Supplier Number:  92-468
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    Semaphorin-4C|Semacl1|Semaphorin I|M-Sema F|S4c|Semai|Sema4c|Sema I|Semaphorin-C-like 1
  • Protein/Peptide Name:
    SEMA4C
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    99.3 kD
  • Sequence:
    Ala21-Gly664
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. No.:
    75791-506
  • Supplier no.:
    92-468

Specifications

About this item

Semaphorin-4C is a protein which belongs to the semaphorin family, contains 1 Ig-like C2-type domain, 1 PSI domain, 1 Sema domain. As cell surface receptor for PLXNB2, it plays an important role in cell-cell signaling. PLXNB2 binding promotes downstream activation of RHOA and phosphorylation of ERBB2 at 'Tyr-1248'. It required for normal brain development, axon guidance and cell migration, Probable signaling receptor which may play a role in myogenic differentiation through activation of the stress-activated MAPK cascade.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.