Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Siglec-3
Mouse Recombinant Siglec-3
Catalog # 75791-394
Supplier:  Prosci
CAS Number:  
Mouse Recombinant Siglec-3
Catalog # 75791-394
Supplier:  Prosci
Supplier Number:  92-412
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    CD33|CD33/Myeloid cell surface antigen CD33/Sialic acid-binding Ig-like lectin 3/Siglec-3/Siglec3
  • Protein/Peptide Name:
    Siglec-3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    25.7 kD
  • Sequence:
    Asp18-Glu240
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75791-394
  • Supplier no.:
    92-412

Specifications

About this item

Mouse myeloid cell surface antigen CD33(CD33) is a member of the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. CD33 contains one Ig-like C2-type domain and one Ig-like V-type domain. CD33 is a putative adhesion molecule of myelomonocytic-derived cells that mediates sialic-acid dependent binding to cells. CD33 preferentially binds to alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, CD33 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. CD33 induces apoptosis in acute myeloid leukemia. CD33 is becoming increasingly important as a target of antibody-mediated therapy in acute myeloid leukaemia (AML).

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.