Order Entry
United States
ContactUsLinkComponent
Human Recombinant Myelin P0-like 1 (from Cells)
Human Recombinant Myelin P0-like 1 (from Cells)
Catalog # 75788-708
Supplier:  Prosci
CAS Number:  
Human Recombinant Myelin P0-like 1 (from Cells)
Catalog # 75788-708
Supplier:  Prosci
Supplier Number:  92-390
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    transcript variant 1|isoform CRA_b|highly similar to Homo sapiens myelin protein zero-like 1 (MPZL1)|cDNA FLJ78597|FLJ96614|Homo sapiens myelin protein zero-like 1 (MPZL1)|Mrna|Myelin protein zero-like 1|MPZL1|cDNA|mRNA
  • Protein/Peptide Name:
    Myelin P0-like 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    15.2 kD
  • Sequence:
    Ser36-Val162
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75788-708
  • Supplier no.:
    92-390

Specifications

About this item

Myelin protein zero-like protein 1(MPZL1) is encoded by the MPZL1 gene, which is a single-pass type I membrane protein. It is widely expressed with highest levels in heart, placenta, kidney and pancreas. As cell surface receptor, it involved in signal transduction processes. MPZL1 recruits PTPN11/SHP-2 to the cell membrane and subsequently activate/phosphorylate Src kinase at Tyr426, promoting phosphorylation of cortactin and migration of HCC cells. MPZL1also is a major receptor for concanavalin-A (ConA) and involved in cellular signaling induced by ConA, which probably includes Src family tyrosine-protein kinases.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.