Order Entry
United States
ContactUsLinkComponent
E. coli Recombinant beta-Glucuronidase (from E. coli)
E. coli Recombinant beta-Glucuronidase (from E. coli)
Catalog # 75791-186
Supplier:  Prosci
CAS Number:  
E. coli Recombinant beta-Glucuronidase (from E. coli)
Catalog # 75791-186
Supplier:  Prosci
Supplier Number:  92-300
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    E. coli
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage Conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme Name:
    beta-Glucuronidase
  • Enzyme Synonyms:
    Beta-glucuronidase, uidA, GUS, Beta-GUS, GUSB
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    69.9 kD
  • Sequence:
    Met1-Gln603
  • Formulation:
    Supplied as a 0.2 µm filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Biological assays
  • Cat. No.:
    75791-186
  • Supplier no.:
    92-300

Specifications

About this item

Beta-D-glucuronidase from E

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

coli is a  highly specific enzyme in hydrolyzing glucuronides. Beta-D-glucuronidase (GLUase) activity can be measured as the rate of production of fluorescent methylumbelliferone (MU), resulting from the hydrolysis of the substrate 4-methylumbelliferyl- beta -d-glucuronide (MUGLU), which is an effective and rapid method for detection and verification of E. coli in food, water, and environmental samples. High purity recombinant beta-Glucuronidase is used in research, biochemical enzyme assays and in vitro diagnostic analysis, detecting a wide range of drugs such as opioids, benzodiazepines, steroids, cannabinoids, and others.