Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant EGF (from E. coli)
Mouse Recombinant EGF (from E. coli)
Catalog # 75791-084
Supplier:  Prosci
CAS Number:  
Mouse Recombinant EGF (from E. coli)
Catalog # 75791-084
Supplier:  Prosci
Supplier Number:  92-245
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    EGF|Pro-epidermal growth factor|Epidermal growth factor
  • Protein/Peptide Name:
    EGF
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    7.2 kD
  • Sequence:
    Asn977-Arg1029
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75791-084
  • Supplier no.:
    92-245

Specifications

About this item

EGF is a single-pass type I membrane protein,containing 8 LDL-receptor class B repeats and 9 EGF-like domains. EGF results in cellular proliferation, differentiation, and survival.EGF is a low-molecular-weight polypeptide first purified from the mouse submandibular gland, but since then found in many human tissues including submandibular gland, parotid gland. Salivary EGF, which seems also regulated by dietary inorganic iodine, also plays an important physiological role in the maintenance of oro-esophageal and gastric tissue integrity. The biological effects of salivary EGF include healing of oral and gastroesophageal ulcers, inhibition of gastric acid secretion, stimulation of DNA synthesis as well as mucosal protection from intraluminal injurious factors such as gastric acid, bile acids, pepsin, and trypsin and to physical, chemical and bacterial agents.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.