Human Recombinant Parvulin-14 (from E. coli)
Catalog # 75790-940
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-6 His tag
- Storage Conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:Peptidyl-prolyl cis/trans isomerase EPVH|PIN4|Parvulin-17|Peptidyl-prolyl cis-trans isomerase Pin4|Parvulin-14|Rotamase Pin4|Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4
- Protein/Peptide Name:Parvulin-14
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:18.8 kD
- Sequence:Met1-Lys156
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75790-940
- Supplier no.:92-173
Specifications
About this item
Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4(PIN4) is a peptidyl-prolyl cis/trans isomerase (PPIase) which interacts with NIMA and is vital for cell cycle regulation. PIN4 has 2 different isoforms: PAR14 and PAR17. Furthermore, PIN4 protein binds to double-stranded DNA under physiological salt conditions. PIN4 is involved as a ribosomal RNA processing factor in ribosome biogenesis. The PAR14 binds to tightly bent AT-rich stretches of double-stranded DNA, but PAR17 binds to double-stranded DNA.
- Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.