Human Recombinant Lysophospholipase II (from E. coli)
Catalog # 75790-786
Supplier: Prosci
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme Name:Lysophospholipase II
- Enzyme Synonyms:Acyl-Protein Thioesterase 2, APT-2, Lysophospholipase II, LPL-II, LysoPLA II, LYPLA2, APT2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular Weight:25.8 kD
- Sequence:Met1-Val231
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM Tris, 1 mM DTT, 0.1 M NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75790-786
- Supplier no.:92-096
Specifications
About this item
Acyl-Protein Thioesterase 2 is a cytoplasmic protein that belongs to the AB Hydrolase 2 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
LYPLA2 is a lysophospholipase and hydrolyzes fatty acids from S-acylated cysteine residues in proteins such as trimeric G alpha proteins or HRAS. LYPLA2 has hydrolase activity that converts Palmitoyl-protein to palmitate and protein. LYPLA2 regulates the multifunctional lysophospholipids by acting on biological membranes. LYPLA2 participates in Glycerophospholipid metabolism pathway, fatty acid metabolic process and lipid metabolic process.