Human Recombinant Annexin A10 (from E. coli)
75790-718
: Prosci
:
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:Annexin-10|ANXA10|ANX14|Annexin-14|Annexin A10
- Protein/Peptide Name:Annexin A10
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:37.5 kD
- Sequence:Met1-Tyr324
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:used for biological assays
- Cat. No.:75790-718
- Supplier no.:92-062
Annexin A10 (ANXA10) contains four Annexin repeats and is a member of the Annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. It is reported that ANXA10 may be a clinical relevant marker for predicting outcome in both early and advanced stages of bladder cancer.
This recombinant protein can be used for biological assays. For research use only.