Human Recombinant BCL-2-associated Athanogene 1 (from E. coli)
Catalog # 75788-684
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:N-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:BAG2|BAG-2|BAG Family Molecular Chaperone Regulator 2|Bcl-2-Associated Athanogene 2
- Protein/Peptide Name:BCL-2-associated Athanogene 1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:25.9 kD
- Sequence:Met1-Asn211
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75788-684
- Supplier no.:92-006
Specifications
About this item
BAG Family Molecular Chaperone Regulator 2 (BAG2) is a member of the Bag family whose members compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. BAG2 contains 1 BAG domain and is a important component of the HSC 70/CHIP chaperone-dependent ubiquitin ligase complex. In mammalian cells BAG1, BAG2, and BAG3 bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner.
- Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.