Human Recombinant Partner of Y14 and Mago (from E. coli)
Catalog # 75788-662
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:Partner of Y14 and Mago|PYM|WIBG|Protein Wibg Homolog
- Protein/Peptide Name:Partner of Y14 and Mago
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:23.7 kD
- Sequence:Met1-Leu204
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75788-662
- Supplier no.:91-994
Specifications
About this item
Partner of Y14 and Mago (WIBG) is a key regulator of the Exon Junction Complex (EJC). EJC is a multiprotein complex that associates immediately upstream of the exon-exon junction on mRNAs, is a positional landmarker for the intron exon structure of genes, and directs post-transcriptional processes in the cytoplasm, for instance mRNA export, nonsense-mediated mRNA decay or translation. WIBG is a cytoplasmic RNA-binding protein, it can be excluded from nucleus by Crm1. WIBG as a cooperateing partner of Mago-14, relates with Mago-14 by its N-terminal domain.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.