Human Recombinant TMIGD2 (from Cells)
Catalog # 75790-450
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:Transmembrane and immunoglobulin domain-containing protein 2|Immunoglobulin and proline-rich receptor 1|TMIGD2|IGPR1|CD28H|IGPR-1
- Protein/Peptide Name:TMIGD2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:17.3 kD
- Sequence:Leu23-Gly150
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:used for biological assays
- Cat. No.:75790-450
- Supplier no.:91-907
Specifications
About this item
TMIGD2 is a single-pass type I membrane protein, which contains one Ig-like (immunoglobulin-like) domain. It is widely expressed in many tissues, such as epithelial, endothelial cells and lung. However, it isn’t detected in thyroid, cerebellum, thymus and cerebral cortex. TMIGD2 can form homophilic interactions that could regulate cell-cell interaction. It Interacts with CACNB2, DST, MIA and NCKIPSD. It is shown that TMIGD2 plays a role in cell-cell interaction, cell migration, and angiogenesis.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.