Human Recombinant Non-T cell activation linker (from Cells)
75790-146
: Prosci
:
- Protein/Peptide Type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:WBSCR5|WBS15|Non-T-Cell Activation Linker|Linker for Activation of T-Cells Family Member 2|Linker for Activation of B-Cells|Membrane-Associated Adapter Molecule|NTAL|LAT2|WBSCR15|LAB|Williams-Beuren Syndrome Chromosomal Region 15 Protein|Williams-Beuren Syndrome Chromosomal Region 5 Protein
- Protein/Peptide Name:Non-T cell activation linker
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:25 kD
- Sequence:Arg27-Ala243
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75790-146
- Supplier no.:91-747
Linker for Activation of T-Cells Family Member 2 (LAT2) is a single-pass type III membrane protein. LAT2 is highly expressed in the spleen, peripheral blood lymphocytes, and germinal centers of lymph nodes. LAT2 is involved in FCER1 (high affinity immunoglobulin epsilon receptor)-mediated signaling in mast cells. It may also be involved in BCR (B-cell antigen receptor)-mediated signaling in B-cells and FCGR1 (high affinity immunoglobulin gamma Fc receptor I)-mediated signaling in myeloid cells. Coupleing activate of these receptors and their associated kinases with distal intracellular events through the recruitment of GRB2.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.