Human Recombinant PRPS2 (from Cells)
75789-950
: Prosci
:
- Protein/Peptide Type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:Ribose-Phosphate Pyrophosphokinase 2|PRPS2|Phosphoribosyl Pyrophosphate Synthase II|PPRibP|PRS-II
- Protein/Peptide Name:PRPS2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:35.8 kD
- Sequence:Pro2-Leu318
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75789-950
- Supplier no.:91-645
Ribose-Phosphate Pyrophosphokinase 2 (PRPS2) is a phosphoribosyl pyrophosphate synthetase that belongs to the ribose-phosphate pyrophosphokinase family. PRPS2 is a homodimer. The active form is probably an hexamer composed of three homodimers. PRPS2 catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis. PRPS2 catalyzes the synthesis of 5-phosphoribosyl 1-pyrophosphate from ATP and D-ribose 5-phosphate. In addition, PRPS2 plays a central role in the synthesis of purines and pyrimidines.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.